Assay Details
| Gene Symbol: HCFC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013320.2 | NP_037452.1 | 11 - 12 | 1767 | 134 | |
| XM_017019241.1 | XP_016874730.1 | 10 - 11 | 1599 | 134 | ||
| XM_017019242.1 | XP_016874731.1 | 11 - 12 | 1767 | 134 | ||
| GenBank mRNA | AF117210.1 | - | 11 - 12 | 1676 | 134 | |
| AK223342.1 | - | 11 - 12 | 1694 | 134 | ||
| AK313466.1 | - | 11 - 12 | 1697 | 134 | ||
| BC006558.2 | - | 10 - 11 | 1630 | 2823984 | 134 | |
| BC033799.1 | - | 11 - 12 | 1669 | 5172046 | 134 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000807.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010271.hg.1 | ENST00000229330.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010271.hg.1 | ENST00000550335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010271.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12010271.hg.1 | TC12000807.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000547194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000550444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | ENST00000552343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008671.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | ENST00000229330.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | ENST00000544223.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | ENST00000550335.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | NM_013320.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200164020.hg.1 | TC1200008671.hg.1 | Clariom D TC |

HCFC2
Genomic Map