Assay Details
| Gene Symbol: NDUFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004547.5 | NP_004538.2 | 2 - 3 | 402 | 102 | |
| GenBank mRNA | AF044957.1 | - | 2 - 3 | 331 | 102 | |
| BC000855.1 | - | 2 - 3 | 374 | 3459215 | 102 | |
| BC070285.1 | - | 2 - 3 | 341 | 6714676 | 102 | |
| BC146885.1 | - | 2 - 3 | 373 | 9056813 | 102 | |
| BC146887.1 | - | 2 - 3 | 373 | 9056815 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000617.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03011164.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011164.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011164.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011164.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011164.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011164.hg.1 | TC03000617.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008383.hg.1 | ENST00000560510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008383.hg.1 | ENST00000560510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008383.hg.1 | ENST00000560510.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008383.hg.1 | ENST00000600530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008383.hg.1 | ENST00000600530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008383.hg.1 | ENST00000600530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011167.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011167.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011167.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03011167.hg.1 | TC03000617.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160610.hg.1 | TC0300008514.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000461682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008514.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160606.hg.1 | TC0300008514.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400008383.hg.1 | ENST00000560510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008383.hg.1 | ENST00000560510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008383.hg.1 | ENST00000560510.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008383.hg.1 | ENST00000600530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008383.hg.1 | ENST00000600530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008383.hg.1 | ENST00000600530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000184266.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000485064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000491335.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000492739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | ENST00000496588.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | NM_001168331.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | NM_004547.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300160607.hg.1 | TC0300008514.hg.1 | Clariom D TC |

NDUFB4
Genomic Map