Assay Details
| Gene Symbol: PGM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172818.1 | NP_001166289.1 | 9 - 10 | 1759 | 86 | |
| NM_001172819.1 | NP_001166290.1 | 9 - 10 | 1550 | 86 | ||
| NM_002633.2 | NP_002624.2 | 9 - 10 | 1673 | 86 | ||
| GenBank mRNA | AK293298.1 | - | 9 - 10 | 1550 | 86 | |
| AK294186.1 | - | 10 - 11 | 1632 | 86 | ||
| AK298505.1 | - | 9 - 10 | 1480 | 86 | ||
| AK300100.1 | - | 9 - 10 | 1745 | 86 | ||
| AK312254.1 | - | 9 - 10 | 1538 | 86 | ||
| BC001756.2 | - | 9 - 10 | 1586 | 3505510 | 86 | |
| BC019920.1 | - | 9 - 10 | 1673 | 5122261 | 86 | |
| BC067763.1 | - | 9 - 10 | 1551 | 5286530 | 86 | |
| BC090856.1 | - | 9 - 10 | 1518 | 86 | ||
| BT006961.1 | - | 9 - 10 | 1460 | 86 | ||
| M83088.1 | - | 9 - 10 | 1522 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000722.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | ENST00000371084.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | ENST00000538673.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | ENST00000540265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011472.hg.1 | TC01000722.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160951.hg.1 | TC0100008591.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371083.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000371084.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000473117.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000483707.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | ENST00000540265.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172818.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_001172819.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008591.hg.1 | NM_002633.2 | Refseq |

PGM1
Genomic Map