Assay Details
| Gene Symbol: NLRC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032206.4 | NP_115582.4 | 20 - 21 | 3374 | 95 | |
| XM_005256193.2 | XP_005256250.1 | 21 - 22 | 3435 | 95 | ||
| XM_005256194.3 | XP_005256251.1 | 21 - 22 | 3646 | 95 | ||
| XM_005256195.2 | XP_005256252.1 | 21 - 22 | 3435 | 95 | ||
| XM_005256196.2 | XP_005256253.1 | 20 - 21 | 3351 | 95 | ||
| XM_005256197.2 | XP_005256254.1 | 21 - 22 | 3435 | 95 | ||
| XM_005256199.3 | XP_005256256.1 | 21 - 22 | 3435 | 95 | ||
| XM_006721297.2 | XP_006721360.1 | 21 - 22 | 3423 | 95 | ||
| XM_006721300.2 | XP_006721363.1 | 21 - 22 | 3435 | 95 | ||
| XM_011523373.2 | XP_011521675.1 | 21 - 22 | 3582 | 95 | ||
| XM_011523375.2 | XP_011521677.1 | 21 - 22 | 3435 | 95 | ||
| XM_011523376.2 | XP_011521678.1 | 16 - 17 | 1261 | 95 | ||
| XM_017023766.1 | XP_016879255.1 | 21 - 22 | 3435 | 95 | ||
| XM_017023767.1 | XP_016879256.1 | 21 - 22 | 3423 | 95 | ||
| XM_017023768.1 | XP_016879257.1 | 21 - 22 | 3435 | 95 | ||
| XM_017023769.1 | XP_016879258.1 | 21 - 22 | 3423 | 95 | ||
| XM_017023770.1 | XP_016879259.1 | 21 - 22 | 3435 | 95 | ||
| XM_017023771.1 | XP_016879260.1 | 21 - 22 | 3435 | 95 | ||
| GenBank mRNA | AF389420.1 | - | 19 - 20 | 3247 | 95 | |
| AK025362.1 | - | 16 - 17 | 2506 | 95 | ||
| AK074133.1 | - | 9 - 10 | 700 | 95 | ||
| AK074182.1 | - | 21 - 22 | 3870 | 95 | ||
| AK090439.1 | - | 16 - 17 | 2429 | 95 | ||
| BC050527.1 | - | 5 - 6 | 339 | 95 | ||
| BC063566.1 | - | 3 - 4 | 160 | 95 | ||
| EF452236.1 | - | 19 - 20 | 3247 | 95 | ||
More Information
Set Membership: |
|

NLRC5
Genomic Map