Assay Details
| Gene Symbol: NLRC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011523375.2 | XP_011521677.1 | 32 - 33 | 4335 | 99 | |
| XM_017023771.1 | XP_016879260.1 | 32 - 33 | 4332 | 99 | ||
More Information
Set Membership: |
|

NLRC5
Genomic Map