Assay Details
| Gene Symbol: KLHL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039548.2 | NP_001034637.2 | 1 - 2 | 883 | 85 | |
| XM_011544954.2 | XP_011543256.1 | 2 - 3 | 2392 | 85 | ||
| GenBank mRNA | BC042952.2 | - | 1 - 2 | 424 | 5275230 | 85 |
| BC057763.1 | - | 1 - 2 | 445 | 85 | ||
| BC132708.1 | - | 1 - 2 | 247 | 40146730 | 85 | |
| BC132710.1 | - | 1 - 2 | 247 | 40146732 | 85 | |
| BI464945.1 | - | 1 - 2 | 450 | 85 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002094.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11025100.hg.1 | ENST00000376292.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025100.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025100.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025100.hg.1 | TC11002094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025098.hg.1 | ENST00000376292.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025098.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025098.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025098.hg.1 | TC11002094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025102.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025102.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025102.hg.1 | TC11002094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025096.hg.1 | ENST00000376292.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025096.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025096.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025096.hg.1 | TC11002094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025099.hg.1 | ENST00000376292.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025099.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025099.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025099.hg.1 | TC11002094.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100189944.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189944.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189944.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189944.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189944.hg.1 | TC1100011651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189941.hg.1 | TC1100011651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189942.hg.1 | TC1100011651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189938.hg.1 | TC1100011651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189940.hg.1 | TC1100011651.hg.1 | Clariom D TC |

KLHL35
Genomic Map