Assay Details
| Gene Symbol: KLHL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039548.2 | NP_001034637.2 | 3 - 4 | 1189 | 65 | |
| XM_011544954.2 | XP_011543256.1 | 4 - 5 | 2698 | 65 | ||
| GenBank mRNA | BC042952.2 | - | 3 - 4 | 730 | 5275230 | 65 |
| BC057763.1 | - | 3 - 4 | 751 | 65 | ||
| BC132708.1 | - | 3 - 4 | 553 | 40146730 | 65 | |
| BC132710.1 | - | 3 - 4 | 553 | 40146732 | 65 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002094.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11025093.hg.1 | ENST00000376292.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025093.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025093.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11025093.hg.1 | TC11002094.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | ENST00000624466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011651.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000527491.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189933.hg.1 | TC1100011651.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | ENST00000376292.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | ENST00000460787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | ENST00000539798.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | NM_001039548.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100189932.hg.1 | TC1100011651.hg.1 | Clariom D TC |

KLHL35
Genomic Map