Assay Details
| Gene Symbol: CYP26A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057157.2 | NP_476498.1 | 1 - 2 | 363 | 88 | |
| GenBank mRNA | AK027560.1 | - | 1 - 2 | 364 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000663.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK027560.1,NM_057157.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
vitamin metabolic process
xenobiotic metabolic process
retinoic acid catabolic process
retinoic acid metabolic process
negative regulation of retinoic acid receptor signaling pathway
oxidation-reduction process
retinoic acid binding
iron ion binding
retinoic acid 4-hydroxylase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
oxygen binding
heme binding
vitamin metabolic process
xenobiotic metabolic process
retinoic acid catabolic process
retinoic acid metabolic process
negative regulation of retinoic acid receptor signaling pathway
oxidation-reduction process
retinoic acid binding
iron ion binding
retinoic acid 4-hydroxylase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
oxygen binding
heme binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000625202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000625202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | ENST00000625202.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007465.hg.1 | ENST00000371531.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007465.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10007465.hg.1 | TC10000663.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1000157625.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000157625.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157625.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000157625.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000157625.hg.1 | TC1000008478.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000224356.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000371531.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000622925.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000623162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000624589.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000625202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000625202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | ENST00000625202.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_000783.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000008478.hg.1 | NM_057157.2 | Refseq |

CYP26A1
Genomic Map