Assay Details
| Gene Symbol: SERPINA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000354.5 | NP_000345.2 | 2 - 3 | 668 | 93 | |
| XM_005262180.3 | XP_005262237.1 | 2 - 3 | 666 | 93 | ||
| XM_006724683.2 | XP_006724746.1 | 2 - 3 | 666 | 93 | ||
| GenBank mRNA | AK310778.1 | - | 2 - 3 | 666 | 93 | |
| BC020747.2 | - | 1 - 2 | 637 | 4716693 | 93 | |
More Information
Set Membership: |
|

SERPINA7
Genomic Map