| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000327674.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000372563.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
ENST00000487487.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010452.hg.1 |
NM_000354.5 |
Refseq |