Assay Details
| Gene Symbol: DIS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322349.1 | NP_001309278.1 | 3 - 4 | 743 | 114 | |
| NM_014953.4 | NP_055768.3 | 2 - 3 | 675 | 114 | ||
| GenBank mRNA | AB001743.1 | - | 2 - 3 | 406 | 114 | |
| AF330044.1 | - | 2 - 3 | 415 | 114 | ||
| AK314715.1 | - | 2 - 3 | 491 | 114 | ||
| AL832266.1 | - | 2 - 3 | 468 | 114 | ||
| BC038101.1 | - | 3 - 4 | 714 | 4827328 | 114 | |
| BC056143.1 | - | 2 - 3 | 480 | 5505671 | 114 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC038101.1,NM_001322349.1 transcripts.
Set Membership: |
|

DIS3
Genomic Map