Assay Details
| Gene Symbol: CD300A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256841.1 | NP_001243770.1 | 3 - 4 | 614 | 110 | |
| NM_007261.3 | NP_009192.2 | 4 - 5 | 953 | 110 | ||
| XM_005256992.4 | XP_005257049.1 | 4 - 5 | 882 | 110 | ||
| XM_006721656.3 | XP_006721719.1 | 4 - 5 | 990 | 110 | ||
| GenBank mRNA | AF020314.1 | - | 5 | 718 | 119 | |
| AJ010101.1 | - | 4 - 5 | 661 | 110 | ||
| AJ010102.1 | - | 3 - 4 | 322 | 110 | ||
| AJ224864.1 | - | 4 - 5 | 934 | 110 | ||
| AJ238323.1 | - | 4 - 5 | 661 | 110 | ||
| AK309631.1 | - | 5 - 6 | 1018 | 110 | ||
| BC032352.1 | - | 4 - 5 | 918 | 5218501 | 110 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000831.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
immune system process
cell adhesion
signal transduction
negative regulation of B cell proliferation
positive regulation of phosphoprotein phosphatase activity
negative regulation of mast cell activation involved in immune response
negative regulation of mast cell degranulation
negative regulation of MAP kinase activity
negative regulation of fibroblast proliferation
regulation of immune response
regulation of T cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of NK T cell activation
negative regulation of phagocytosis, engulfment
negative regulation of neutrophil activation
negative regulation of eosinophil activation
negative regulation of activation of JAK2 kinase activity
negative regulation of eosinophil migration
phosphatidylserine binding
protein binding
phosphatidylethanolamine binding
signaling receptor activity
immune system process
cell adhesion
signal transduction
negative regulation of B cell proliferation
positive regulation of phosphoprotein phosphatase activity
negative regulation of mast cell activation involved in immune response
negative regulation of mast cell degranulation
negative regulation of MAP kinase activity
negative regulation of fibroblast proliferation
regulation of immune response
regulation of T cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of NK T cell activation
negative regulation of phagocytosis, engulfment
negative regulation of neutrophil activation
negative regulation of eosinophil activation
negative regulation of activation of JAK2 kinase activity
negative regulation of eosinophil migration
phosphatidylserine binding
protein binding
phosphatidylethanolamine binding
signaling receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17010740.hg.1 | ENST00000310828.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010740.hg.1 | ENST00000360141.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010740.hg.1 | ENST00000361933.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010740.hg.1 | ENST00000392625.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010740.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17010740.hg.1 | TC17000831.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008852.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | NM_007261.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | TC1700008852.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000310828.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000360141.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000361933.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000392625.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000577511.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | ENST00000582319.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | NM_001256841.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700166127.hg.1 | NM_007261.3 | Refseq |

CD300A
Genomic Map