| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000330767.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000389982.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
ENST00000576060.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700006742.hg.1 |
NM_198154.2 |
Refseq |