Assay Details
| Gene Symbol: TMEM95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320435.1 | NP_001307364.1 | 3 - 4 | 375 | 89 | |
| NM_001320436.1 | NP_001307365.1 | 3 - 4 | 375 | 89 | ||
| NM_198154.2 | NP_937797.1 | 3 - 4 | 375 | 113 | ||
| XM_017024565.1 | XP_016880054.1 | 3 - 4 | 397 | 89 | ||
| XM_017024568.1 | XP_016880057.1 | 3 - 4 | 397 | 89 | ||
| XM_017024569.1 | XP_016880058.1 | 3 - 4 | 397 | 89 | ||
| XM_017024571.1 | XP_016880060.1 | 3 - 4 | 543 | 89 | ||
| GenBank mRNA | AY358218.1 | - | 3 - 4 | 333 | 113 | |
| BC040900.1 | - | 3 - 4 | 375 | 89 | ||
| BC107110.2 | - | 3 - 4 | 333 | 40022149 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

TMEM95
Genomic Map