Assay Details
| Gene Symbol: PZP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002864.2 | NP_002855.2 | 5 - 6 | 527 | 69 | |
| XM_011520804.2 | XP_011519106.1 | 5 - 6 | 526 | 69 | ||
| XM_011520805.2 | XP_011519107.1 | 5 - 6 | 526 | 69 | ||
| XM_011520806.1 | XP_011519108.1 | 5 - 6 | 526 | 69 | ||
| XM_011520807.2 | XP_011519109.1 | 5 - 6 | 526 | 69 | ||
| XM_017019763.1 | XP_016875252.1 | 5 - 6 | 526 | 69 | ||
| XM_017019764.1 | XP_016875253.1 | 5 - 6 | 526 | 69 | ||
| XM_017019765.1 | XP_016875254.1 | 5 - 6 | 526 | 69 | ||
| XM_017019766.1 | XP_016875255.1 | 4 - 5 | 675 | 69 | ||
| XM_017019767.1 | XP_016875256.1 | 5 - 6 | 526 | 69 | ||
| GenBank mRNA | AK313639.1 | - | 5 - 6 | 526 | 69 | |
| BC111756.1 | - | 4 - 5 | 675 | 40027384 | 69 | |
| X54380.1 | - | 5 - 6 | 527 | 69 | ||
More Information
Set Membership: |
|

PZP
Genomic Map