Assay Details
| Gene Symbol: KLHL7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006715755.2 | XP_006715818.1 | 1 - 2 | 576 | 68 | |
| XM_017012439.1 | XP_016867928.1 | 1 - 2 | 576 | 68 | ||
| GenBank mRNA | AK302995.1 | - | 1 - 2 | 133 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK302995.1, XM_006715755.2, XM_017012439.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000322275.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000339077.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000409689.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000410047.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000414163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000414163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000414163.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000459661.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469576.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000469845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000477076.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479288.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000479700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000491352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | ENST00000521082.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001031710.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_001172428.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NM_018846.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033328.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700006906.hg.1 | NR_033329.1 | Refseq |

KLHL7
Genomic Map