Assay Details
| Gene Symbol: SPACA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145248.4 | NP_660291.2 | 1 - 2 | 138 | 156 | |
| XM_011537466.1 | XP_011535768.1 | 1 - 2 | 163 | 156 | ||
| GenBank mRNA | AK130866.1 | - | 1 - 2 | 138 | 156 | |
| BC016750.2 | - | 1 - 2 | 120 | 4837062 | 156 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000409.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13004439.hg.1 | ENST00000283550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004439.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004439.hg.1 | TC13000409.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004435.hg.1 | ENST00000283550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004435.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004435.hg.1 | TC13000409.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004436.hg.1 | ENST00000283550.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004436.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13004436.hg.1 | TC13000409.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154297.hg.1 | TC1300008072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154292.hg.1 | TC1300008072.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000375699.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008072.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | ENST00000283550.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | ENST00000414180.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | ENST00000443541.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | NM_145248.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300154294.hg.1 | TC1300008072.hg.1 | Clariom D TC |

SPACA7
Genomic Map