Assay Details
| Gene Symbol: C9orf116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048265.1 | NP_001041730.1 | 2 - 3 | 285 | 104 | |
| GenBank mRNA | AY927868.1 | - | 2 - 3 | 221 | 104 | |
| DR003074.1 | - | 2 - 3 | 368 | 104 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001715.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021910.hg.1 | ENST00000429260.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021910.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09021910.hg.1 | TC09001715.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371789.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000371791.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000419770.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011938.hg.1 | NM_144654.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | ENST00000371786.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | ENST00000429260.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | NM_001048265.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900181608.hg.1 | TC0900011938.hg.1 | Clariom D TC |

C9orf116
Genomic Map