Assay Details
| Gene Symbol: PHF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005392.3 | NP_005383.3 | 8 - 9 | 1183 | 64 | |
| XM_005252051.2 | XP_005252108.1 | 8 - 9 | 1177 | 64 | ||
| XM_006717143.2 | XP_006717206.1 | 8 - 9 | 1072 | 64 | ||
| GenBank mRNA | AB014562.2 | - | 7 - 8 | 1076 | 64 | |
| AL834263.1 | - | 8 - 9 | 1034 | 64 | ||
| BC152437.1 | - | 7 - 8 | 1076 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000450.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
liver development
transcription, DNA-templated
protein demethylation
histone H3-K9 demethylation
oxidation-reduction process
negative regulation of chromatin silencing at rDNA
transcription coactivator activity
iron ion binding
protein binding
zinc ion binding
histone demethylase activity
histone demethylase activity (H3-K9 specific)
methylated histone binding
dioxygenase activity
liver development
transcription, DNA-templated
protein demethylation
histone H3-K9 demethylation
oxidation-reduction process
negative regulation of chromatin silencing at rDNA
transcription coactivator activity
iron ion binding
protein binding
zinc ion binding
histone demethylase activity
histone demethylase activity (H3-K9 specific)
methylated histone binding
dioxygenase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09004858.hg.1 | ENST00000359246.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004858.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004858.hg.1 | TC09000450.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004859.hg.1 | ENST00000359246.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004859.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004859.hg.1 | TC09000450.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000375376.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | ENST00000610682.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007994.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153862.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153862.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153862.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153862.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153862.hg.1 | TC0900007994.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900153861.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153861.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153861.hg.1 | ENST00000359246.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153861.hg.1 | NM_005392.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153861.hg.1 | TC0900007994.hg.1 | Clariom D TC |

PHF2
Genomic Map