Assay Details
| Gene Symbol: GID4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024052.4 | NP_076957.3 | 5 - 6 | 1004 | 85 | |
| GenBank mRNA | AK127580.1 | - | 5 - 6 | 691 | 85 | |
| AK315083.1 | - | 5 - 6 | 892 | 85 | ||
| BC000636.2 | - | 5 - 6 | 665 | 85 | ||
| BC041829.1 | - | 5 - 6 | 1009 | 5268071 | 85 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000203.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000579871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | ENST00000579871.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002848.hg.1 | ENST00000268719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002848.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002848.hg.1 | TC17000203.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002850.hg.1 | ENST00000268719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002850.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17002850.hg.1 | TC17000203.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000376345.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000459843.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000579871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | ENST00000579871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007097.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150002.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150002.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150002.hg.1 | TC1700007097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150002.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150002.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | ENST00000579871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | TC1700007097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | ENST00000579871.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150003.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | TC1700007097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700150000.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | NM_024052.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | TC1700007097.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | ENST00000268719.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | ENST00000481836.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700149999.hg.1 | NM_024052.4 | Refseq |

GID4
Genomic Map