Assay Details
| Gene Symbol: TEX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031276.2 | NP_112566.2 | 2 - 3 | 115 | 127 | |
| XM_011530994.1 | XP_011529296.1 | 2 - 3 | 109 | 127 | ||
| XM_017029649.1 | XP_016885138.1 | 2 - 3 | 109 | 127 | ||
| GenBank mRNA | AK292471.1 | - | 2 - 3 | 109 | 127 | |
| BC036016.1 | - | 2 - 3 | 115 | 5270970 | 127 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK292471.1, BC036016.1, NM_031276.2, XM_011530994.1, XM_017029649.1 transcripts.
Set Membership: |
|

TEX11
Genomic Map