Assay Details
| Gene Symbol: CADPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003716.3 | NP_003707.2 | 18 - 19 | 3069 | 132 | |
| XM_011534180.1 | XP_011532482.1 | 20 - 21 | 3235 | 132 | ||
| XM_011534194.2 | XP_011532496.1 | 19 - 20 | 2721 | 132 | ||
| XM_017007358.1 | XP_016862847.1 | 21 - 22 | 3304 | 132 | ||
| XM_017007361.1 | XP_016862850.1 | 20 - 21 | 3235 | 132 | ||
| XM_017007369.1 | XP_016862858.1 | 19 - 20 | 3076 | 132 | ||
| XM_017007380.1 | XP_016862869.1 | 19 - 20 | 3127 | 132 | ||
| XM_017007384.1 | XP_016862873.1 | 19 - 20 | 3076 | 132 | ||
| GenBank mRNA | AB032947.2 | - | 18 - 19 | 3059 | 132 | |
More Information
Set Membership: |
|

CADPS
Genomic Map