| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022988.hg.1 |
ENST00000296129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022988.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022988.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022988.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022988.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022988.hg.1 |
TC03001345.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022990.hg.1 |
ENST00000296129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022990.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022990.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022990.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022990.hg.1 |
TC03001345.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022991.hg.1 |
ENST00000296129.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022991.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022991.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022991.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03022991.hg.1 |
TC03001345.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300010913.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
ENST00000490471.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179536.hg.1 |
TC0300010913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179538.hg.1 |
TC0300010913.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
ENST00000296129.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
ENST00000425231.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
NM_022842.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
NM_178181.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300179540.hg.1 |
TC0300010913.hg.1 |
Clariom D TC |