Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022842.4 | NP_073753.3 | 6 - 7 | 1767 | 91 | |
| XM_011534024.1 | XP_011532326.1 | 6 - 7 | 1725 | 91 | ||
| XM_017007070.1 | XP_016862559.1 | 6 - 7 | 1753 | 91 | ||
| GenBank mRNA | AF468010.1 | - | 6 - 7 | 1742 | 91 | |
| AK023834.1 | - | 4 - 5 | 1345 | 91 | ||
| AK026187.1 | - | 3 - 4 | 784 | 91 | ||
| AK026622.1 | - | 6 - 7 | 1742 | 91 | ||
| AK314441.1 | - | 6 - 7 | 1731 | 91 | ||
| AY026461.1 | - | 6 - 7 | 1714 | 91 | ||
| AY167484.1 | - | 6 - 7 | 1724 | 91 | ||
| AY375452.1 | - | 6 - 7 | 1713 | 91 | ||
| BC069254.1 | - | 6 - 7 | 1720 | 91 | ||
| KU215672.1 | - | 5 - 6 | 1545 | 91 | ||
| KU257676.1 | - | 5 - 6 | 1542 | 91 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001345.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03022980.hg.1 | ENST00000296129.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022980.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022980.hg.1 | TC03001345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000490471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000490471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | ENST00000490471.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000425231.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000490471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000490471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | ENST00000490471.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010913.hg.1 | NM_178181.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179528.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179528.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179528.hg.1 | ENST00000296129.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179528.hg.1 | NM_022842.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179528.hg.1 | TC0300010913.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |

CDCP1
Genomic Map