Assay Details
| Gene Symbol: DRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145038.4 | NP_659475.2 | 7 - 8 | 960 | 134 | |
| XM_005264637.3 | XP_005264694.1 | 6 - 7 | 768 | 134 | ||
| GenBank mRNA | AK057222.1 | - | 7 - 8 | 956 | 134 | |
| AK289953.1 | - | 7 - 8 | 943 | 134 | ||
| AL833892.1 | - | 6 - 7 | 694 | 134 | ||
| BC009488.2 | - | 6 - 7 | 683 | 134 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000151.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02001788.hg.1 | ENST00000288710.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001788.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001788.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02001788.hg.1 | TC02000151.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000421869.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000439066.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000487307.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | ENST00000497651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007038.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | ENST00000288710.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | ENST00000442810.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | ENST00000483675.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | NM_145038.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200147889.hg.1 | TC0200007038.hg.1 | Clariom D TC |

DRC1
Genomic Map