| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000052754.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000393155.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000420120.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000425043.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000441303.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000547568.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000548218.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000552962.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_001920.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133503.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133504.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133506.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000052754.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000393155.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000420120.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000425043.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000441303.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000547568.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000548218.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
ENST00000552962.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_001920.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133503.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133504.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133505.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133506.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
NM_133507.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200189076.hg.1 |
TC1200011496.hg.1 |
Clariom D TC |