Assay Details
| Gene Symbol: NAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005967.3 | NP_005958.1 | 4 - 5 | 1522 | 61 | |
| XM_005268894.2 | XP_005268951.1 | 4 - 5 | 1522 | 61 | ||
| GenBank mRNA | AK314229.1 | - | 4 - 5 | 1302 | 61 | |
| BC065931.1 | - | 4 - 5 | 1309 | 6157017 | 61 | |
| U48361.1 | - | 4 - 5 | 1301 | 61 | ||
| X70991.1 | - | 3 - 4 | 994 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000520.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
endochondral ossification
transcription, DNA-templated
nervous system development
cell proliferation
Schwann cell differentiation
negative regulation of transcription from RNA polymerase III promoter
myelination
regulation of epidermis development
transcription corepressor activity
protein binding
transcription factor binding
endochondral ossification
transcription, DNA-templated
nervous system development
cell proliferation
Schwann cell differentiation
negative regulation of transcription from RNA polymerase III promoter
myelination
regulation of epidermis development
transcription corepressor activity
protein binding
transcription factor binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006800.hg.1 | ENST00000300131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006800.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006800.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006800.hg.1 | TC12000520.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006803.hg.1 | ENST00000300131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006803.hg.1 | ENST00000357680.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006803.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006803.hg.1 | TC12000520.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006802.hg.1 | ENST00000300131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006802.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006802.hg.1 | ENST00000357680.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006802.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006802.hg.1 | TC12000520.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006799.hg.1 | ENST00000300131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006799.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006799.hg.1 | ENST00000357680.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006799.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006799.hg.1 | TC12000520.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006801.hg.1 | ENST00000300131.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006801.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006801.hg.1 | ENST00000357680.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006801.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12006801.hg.1 | TC12000520.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157115.hg.1 | TC1200007859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157118.hg.1 | TC1200007859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157117.hg.1 | TC1200007859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157119.hg.1 | TC1200007859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157120.hg.1 | TC1200007859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000342556.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554718.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000554839.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | ENST00000555857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007859.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157121.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157121.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157121.hg.1 | ENST00000300131.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200157121.hg.1 | NM_005967.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200157121.hg.1 | TC1200007859.hg.1 | Clariom D TC |

NAB2
Genomic Map