Assay Details
| Gene Symbol: ST6GALNAC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175040.3 | NP_778205.1 | 1 - 2 | 202 | 64 | |
| GenBank mRNA | AK292049.1 | - | 1 - 2 | 193 | 64 | |
| CB126299.1 | - | 1 - 2 | 202 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001615.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292049.1,NM_175040.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
ganglioside biosynthetic process
glycolipid metabolic process
oligosaccharide metabolic process
O-glycan processing
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl-galactosaminide 6-alpha-sialyltransferase activity
ganglioside biosynthetic process
glycolipid metabolic process
oligosaccharide metabolic process
O-glycan processing
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
(alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl-galactosaminide 6-alpha-sialyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | ENST00000335791.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | ENST00000343609.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | ENST00000541933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020361.hg.1 | TC09001615.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000474282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000479747.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011610.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000335791.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000361444.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000467674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000483438.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | ENST00000495983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | NM_175039.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | NM_175040.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179214.hg.1 | TC0900011610.hg.1 | Clariom D TC |

ST6GALNAC4
Genomic Map