Assay Details
| Gene Symbol: GATAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021167.4 | NP_066990.3 | 2 - 3 | 657 | 141 | |
| NR_052016.1 | 2 - 3 | 657 | 141 | |||
| GenBank mRNA | AK026142.1 | - | 2 - 3 | 626 | 141 | |
| AK315788.1 | - | 2 - 3 | 623 | 141 | ||
| BC019350.1 | - | 2 - 3 | 628 | 3626609 | 141 | |
| BC031091.1 | - | 2 - 3 | 531 | 5275963 | 141 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003394.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07007993.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007993.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007993.hg.1 | TC07003394.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007992.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007992.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007992.hg.1 | TC07003394.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000465247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000465247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000465247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007995.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007995.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007995.hg.1 | TC07003394.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007991.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007991.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07007991.hg.1 | TC07003394.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000465247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000465247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000465247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013400.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | ENST00000493878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197719.hg.1 | TC0700013400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197716.hg.1 | TC0700013400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197715.hg.1 | TC0700013400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197714.hg.1 | TC0700013400.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | ENST00000287957.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | NM_021167.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | NR_052016.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197717.hg.1 | TC0700013400.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 183733 | - | - |

GATAD1
Genomic Map