Assay Details
| Gene Symbol: SH3PXD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017995.2 | NP_001017995.1 | 8 - 9 | 836 | 68 | |
| NM_001308175.1 | NP_001295104.1 | 8 - 9 | 836 | 68 | ||
| GenBank mRNA | AB430862.1 | - | 8 - 9 | 665 | 68 | |
| BC038561.1 | - | 8 - 9 | 671 | 68 | ||
| DQ109556.1 | - | 8 - 9 | 671 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05002062.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
eye development
superoxide metabolic process
heart development
extracellular matrix disassembly
cell differentiation
positive regulation of catalytic activity
positive regulation of fat cell differentiation
oxidation-reduction process
bone development
adipose tissue development
podosome assembly
protein localization to membrane
protein binding
phosphatidylinositol-5-phosphate binding
superoxide-generating NADPH oxidase activator activity
phosphatidylinositol-3-phosphate binding
SH2 domain binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
skeletal system development
eye development
superoxide metabolic process
heart development
extracellular matrix disassembly
cell differentiation
positive regulation of catalytic activity
positive regulation of fat cell differentiation
oxidation-reduction process
bone development
adipose tissue development
podosome assembly
protein localization to membrane
protein binding
phosphatidylinositol-5-phosphate binding
superoxide-generating NADPH oxidase activator activity
phosphatidylinositol-3-phosphate binding
SH2 domain binding
phosphatidylinositol-4-phosphate binding
phosphatidylinositol-3,5-bisphosphate binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05028413.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028413.hg.1 | ENST00000519643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028413.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028413.hg.1 | TC05002062.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000518522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000518522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000518522.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000523651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | ENST00000523651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028411.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028411.hg.1 | ENST00000519643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028411.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028411.hg.1 | TC05002062.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028412.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028412.hg.1 | ENST00000519643.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028412.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05028412.hg.1 | TC05002062.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000518522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000518522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000518522.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000523651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | ENST00000523651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012826.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189284.hg.1 | TC0500012826.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189282.hg.1 | TC0500012826.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | ENST00000311601.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | ENST00000519643.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | NM_001017995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | NM_001308175.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500189283.hg.1 | TC0500012826.hg.1 | Clariom D TC |

SH3PXD2B
Genomic Map