| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001651.hg.1 |
ENST00000505624.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001651.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001651.hg.1 |
ENST00000535797.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001651.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05001651.hg.1 |
TC05000153.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500148086.hg.1 |
TC0500007120.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502674.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000502782.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000505624.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000506758.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508377.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000508722.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000514080.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
ENST00000610313.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007120.hg.1 |
NM_144725.3 |
Refseq |