Assay Details
| Gene Symbol: TTC23L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001317949.1 | NP_001304878.1 | 9 - 10 | 1113 | 88 | |
| NM_144725.3 | NP_653326.3 | 9 - 10 | 1178 | 88 | ||
| GenBank mRNA | AK058168.1 | - | 9 - 10 | 1178 | 88 | |
| BC057775.1 | - | 9 - 10 | 1150 | 5295348 | 88 | |
| BX088767.1 | - | 4 - 5 | 455 | 88 | ||
| JN799997.1 | - | 9 - 10 | 1113 | 88 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000153.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001654.hg.1 | ENST00000505624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001654.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05001654.hg.1 | TC05000153.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000502782.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000506758.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508377.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000508722.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000514080.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | ENST00000610313.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007120.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148089.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148089.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148089.hg.1 | ENST00000505624.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500148089.hg.1 | NM_144725.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500148089.hg.1 | TC0500007120.hg.1 | Clariom D TC |

TTC23L
Genomic Map