| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004022.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004022.hg.1 |
ENST00000538599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004022.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004022.hg.1 |
TC20000282.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004021.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004021.hg.1 |
ENST00000538599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004021.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20004021.hg.1 |
TC20000282.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000007313.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150583.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150583.hg.1 |
NM_004139.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000150583.hg.1 |
TC2000007313.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000150583.hg.1 |
ENST00000217407.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000150583.hg.1 |
NM_004139.4 |
Refseq |