Assay Details
| Gene Symbol: LBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004139.4 | NP_004130.2 | 11 - 12 | 1321 | 84 | |
| GenBank mRNA | AF105067.1 | - | 11 - 12 | 1232 | 84 | |
| AK313625.1 | - | 11 - 12 | 1251 | 84 | ||
| BC022256.1 | - | 11 - 12 | 1250 | 4767724 | 84 | |
| BQ028088.1 | - | 4 - 5 | 84 | |||
| M35533.1 | - | 11 - 12 | 1203 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000282.hg.1
Set Membership: |
|

LBP
Genomic Map