| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002447.hg.1 |
ENST00000375921.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002447.hg.1 |
ENST00000375934.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002447.hg.1 |
ENST00000375938.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002447.hg.1 |
ENST00000535842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002447.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002447.hg.1 |
TC20000200.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000310998.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000375921.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000375922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000375934.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000375935.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000375938.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
ENST00000535842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002441.hg.1 |
TC20000200.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
ENST00000375921.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
ENST00000375934.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
ENST00000375935.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
ENST00000375938.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
ENST00000535842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002446.hg.1 |
TC20000200.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
ENST00000375921.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
ENST00000375922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
ENST00000375934.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
ENST00000375938.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
ENST00000535842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002451.hg.1 |
TC20000200.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002452.hg.1 |
ENST00000375921.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002452.hg.1 |
ENST00000375922.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002452.hg.1 |
ENST00000375938.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002452.hg.1 |
ENST00000535842.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002452.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20002452.hg.1 |
TC20000200.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000310998.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375921.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375922.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375934.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375938.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000535842.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000611624.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000612972.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000616761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000619753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000620043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000611624.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
TC2000007094.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000310998.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375921.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375922.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375934.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000375938.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000535842.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000612972.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000616761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000619753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
ENST00000620043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148452.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375921.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375922.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375934.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375938.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000535842.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000611624.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000616761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000619753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000620043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
NM_001008409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
TC2000007094.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375921.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375922.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375934.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000375938.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000535842.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000611624.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000616761.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000619753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
ENST00000620043.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000148455.hg.1 |
NM_001008409.3 |
Refseq |