Assay Details
| Gene Symbol: KCNAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004732.3 | NP_004723.2 | 1 - 2 | 243 | 117 | |
| GenBank mRNA | BC096232.1 | - | 1 - 2 | 243 | 40005293 | 117 |
| BC096234.3 | - | 1 - 2 | 243 | 40005295 | 117 | |
| BC099634.1 | - | 1 - 2 | 243 | 40005292 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001097.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17014770.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014770.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014770.hg.1 | TC17001097.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014773.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014773.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014773.hg.1 | TC17001097.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014772.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014772.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014772.hg.1 | TC17001097.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | TC1700012351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205164.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | TC1700012351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205159.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | TC1700012351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205163.hg.1 | NM_004732.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 154892 | - | - |

KCNAB3
Genomic Map