Assay Details
Gene Symbol: KCNAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004732.3 | NP_004723.2 | 4 - 5 | 404 | 62 | |
XM_011524068.1 | XP_011522370.1 | 4 - 5 | 351 | 62 | ||
XM_017025304.1 | XP_016880793.1 | 4 - 5 | 351 | 62 | ||
XM_017025305.1 | XP_016880794.1 | 4 - 5 | 351 | 62 | ||
XM_017025306.1 | XP_016880795.1 | 4 - 5 | 351 | 62 | ||
GenBank mRNA | AF016411.1 | - | 4 - 5 | 404 | 62 | |
BC096232.1 | - | 4 - 5 | 404 | 40005293 | 62 | |
BC096234.3 | - | 4 - 5 | 404 | 40005295 | 62 | |
BC099634.1 | - | 4 - 5 | 404 | 40005292 | 62 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001097.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17014767.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17014767.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17014767.hg.1 | TC17001097.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700205154.hg.1 | TC1700012351.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 154892 | - | - |
