Assay Details
| Gene Symbol: KCNAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004732.3 | NP_004723.2 | 6 - 7 | 493 | 90 | |
| XM_011524068.1 | XP_011522370.1 | 6 - 7 | 440 | 90 | ||
| XM_017025304.1 | XP_016880793.1 | 6 - 7 | 440 | 90 | ||
| XM_017025305.1 | XP_016880794.1 | 6 - 7 | 440 | 90 | ||
| XM_017025306.1 | XP_016880795.1 | 6 - 7 | 440 | 90 | ||
| GenBank mRNA | AF016411.1 | - | 6 - 7 | 493 | 90 | |
| BC096232.1 | - | 6 - 7 | 493 | 40005293 | 90 | |
| BC096234.3 | - | 6 - 7 | 493 | 40005295 | 90 | |
| BC099634.1 | - | 6 - 7 | 493 | 40005292 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001097.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014765.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014765.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17014765.hg.1 | TC17001097.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000570852.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000572275.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000574006.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700012351.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | NM_004732.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | TC1700012351.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | ENST00000303790.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | ENST00000570587.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | ENST00000576981.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700205152.hg.1 | NM_004732.3 | Refseq |

KCNAB3
Genomic Map