Assay Details
| Gene Symbol: ALPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052947.3 | NP_443179.3 | 10 - 11 | 6237 | 78 | |
| XM_011525801.2 | XP_011524103.1 | 10 - 11 | 6470 | 78 | ||
| XM_011525802.2 | XP_011524104.1 | 8 - 9 | 6857 | 78 | ||
| XM_017025535.1 | XP_016881024.1 | 10 - 11 | 6471 | 78 | ||
| XM_017025536.1 | XP_016881025.1 | 10 - 11 | 6471 | 78 | ||
| XM_017025538.1 | XP_016881027.1 | 7 - 8 | 5355 | 78 | ||
| XM_017025539.1 | XP_016881028.1 | 7 - 8 | 4103 | 78 | ||
| GenBank mRNA | AL832018.2 | - | 7 - 8 | 4569 | 78 | |
| AY044450.1 | - | 7 - 8 | 4304 | 78 | ||
| BX647446.1 | - | 6 - 7 | 3715 | 78 | ||
| BX647639.1 | - | 7 - 8 | 5103 | 78 | ||
| BX647796.1 | - | 10 - 11 | 6224 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000542.hg.1
Set Membership: |
|

ALPK2
Genomic Map