| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004301.hg.1 |
ENST00000434431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004301.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004301.hg.1 |
TC02004962.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016441.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016441.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016441.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016441.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016441.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016441.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004300.hg.1 |
ENST00000434431.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004300.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02004300.hg.1 |
TC02004962.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0200221672.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200221672.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200221672.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200221672.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200221672.hg.1 |
TC0200016441.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200016441.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016441.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016441.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016441.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016441.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016441.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200221671.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200221671.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200221671.hg.1 |
ENST00000434431.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200221671.hg.1 |
NM_001145051.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200221671.hg.1 |
TC0200016441.hg.1 |
Clariom D TC |