| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037092.hg.1 |
ENST00000357582.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037092.hg.1 |
ENST00000374040.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037092.hg.1 |
ENST00000493901.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037092.hg.1 |
ENST00000545447.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037092.hg.1 |
NM_004672.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037092.hg.1 |
TC01002405.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000357582.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000374040.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000472410.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000493901.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
NM_001297609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
NM_004672.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000357582.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000374040.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000472410.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
ENST00000493901.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
NM_001297609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
NM_004672.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199740.hg.1 |
TC0100013428.hg.1 |
Clariom D TC |