| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002332.hg.1 |
ENST00000328024.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002332.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002332.hg.1 |
TC19000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002333.hg.1 |
ENST00000328024.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002333.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002333.hg.1 |
TC19000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002330.hg.1 |
ENST00000328024.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002330.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002330.hg.1 |
TC19000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002331.hg.1 |
ENST00000328024.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002331.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19002331.hg.1 |
TC19000149.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900006893.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
TC1900006893.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148874.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
TC1900006893.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
ENST00000601897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148879.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148868.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148868.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148868.hg.1 |
TC1900006893.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900148868.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148868.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
TC1900006893.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148869.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
NM_004218.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
TC1900006893.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000328024.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000594216.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000598706.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
ENST00000600719.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900148876.hg.1 |
NM_004218.3 |
Refseq |