Assay Details
| Gene Symbol: MTERF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033050.2 | NP_001028222.1 | 2 - 3 | 304 | 137 | |
| XM_011538767.2 | XP_011537069.1 | 2 - 3 | 1387 | 137 | ||
| GenBank mRNA | AK024124.1 | - | 2 - 3 | 289 | 137 | |
| AL832827.1 | - | 1 - 2 | 202 | 137 | ||
| BC036066.1 | - | 2 - 3 | 1927 | 5313686 | 137 | |
| DA074327.1 | - | 2 - 3 | 304 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK024124.1, AL832827.1, BC036066.1, NM_001033050.2, XM_011538767.2 transcripts.
Set Membership: |
|

MTERF2
Genomic Map