| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017986.hg.1 |
ENST00000318586.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017986.hg.1 |
ENST00000388914.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017986.hg.1 |
NM_001042784.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04017986.hg.1 |
TC04001309.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
ENST00000388914.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
ENST00000504667.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
NM_001042784.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
ENST00000388914.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
ENST00000504667.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
NM_001042784.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400175429.hg.1 |
TC0400011061.hg.1 |
Clariom D TC |