Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015596.1 | NP_056411.1 | 2 - 3 | 281 | 57 | |
| GenBank mRNA | AB108824.1 | - | 2 - 3 | 238 | 57 | |
| AL050220.1 | - | 2 - 3 | 286 | 57 | ||
| AY923171.1 | - | 2 - 3 | 281 | 57 | ||
| AY923172.1 | - | 3 - 4 | 557 | 57 | ||
| AY923173.1 | - | 3 - 4 | 557 | 57 | ||
| AY923174.1 | - | 2 - 3 | 473 | 57 | ||
| AY923175.1 | - | 2 - 3 | 473 | 57 | ||
| BC069334.1 | - | 2 - 3 | 282 | 7262205 | 57 | |
| BC069543.1 | - | 2 - 3 | 281 | 7262217 | 57 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001770.hg.1
Set Membership: |
|

KLK13
Genomic Map