Assay Details
| Gene Symbol: ZPLD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175056.1 | NP_778226.1 | 6 - 7 | 831 | 141 | |
| XM_017005703.1 | XP_016861192.1 | 7 - 8 | 1013 | 141 | ||
| XM_017005704.1 | XP_016861193.1 | 6 - 7 | 869 | 141 | ||
| GenBank mRNA | AY090780.1 | - | 6 - 7 | 831 | 141 | |
| BC031261.1 | - | 13 - 14 | 1565 | 5273136 | 141 | |
| BC143817.1 | - | 7 - 8 | 927 | 141 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000528.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03009819.hg.1 | ENST00000306176.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009819.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009819.hg.1 | ENST00000491959.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009819.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03009819.hg.1 | TC03000528.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008196.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | ENST00000306176.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | ENST00000466937.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | ENST00000491959.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | NM_175056.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300158396.hg.1 | TC0300008196.hg.1 | Clariom D TC |

ZPLD1
Genomic Map