Assay Details
| Gene Symbol: CPSF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321833.1 | NP_001308762.1 | 11 - 12 | 1862 | 94 | |
| NM_001321834.1 | NP_001308763.1 | 11 - 12 | 2770 | 94 | ||
| NM_001321835.1 | NP_001308764.1 | 11 - 12 | 1505 | 94 | ||
| NM_001321836.1 | NP_001308765.1 | 12 - 13 | 1952 | 94 | ||
| NM_016207.3 | NP_057291.1 | 11 - 12 | 1633 | 94 | ||
| GenBank mRNA | AF017269.1 | - | 7 - 8 | 1039 | 94 | |
| AF171877.1 | - | 11 - 12 | 1411 | 94 | ||
| AK025440.1 | - | 9 - 10 | 1292 | 94 | ||
| AK223487.1 | - | 11 - 12 | 1431 | 94 | ||
| AK298918.1 | - | 9 - 10 | 1226 | 94 | ||
| BC011654.2 | - | 11 - 12 | 1506 | 4304135 | 94 | |
| BC014106.1 | - | 11 - 12 | 2423 | 94 | ||
| BC020211.1 | - | 11 - 12 | 1431 | 4890593 | 94 | |
| BC030988.1 | - | 10 - 11 | 1372 | 94 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000047.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA polyadenylation
mRNA cleavage
mRNA 3'-end processing by stem-loop binding and cleavage
mRNA export from nucleus
mRNA 3'-end processing
RNA phosphodiester bond hydrolysis, endonucleolytic
RNA binding
endoribonuclease activity
protein binding
5'-3' exonuclease activity
metal ion binding
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA polyadenylation
mRNA cleavage
mRNA 3'-end processing by stem-loop binding and cleavage
mRNA export from nucleus
mRNA 3'-end processing
RNA phosphodiester bond hydrolysis, endonucleolytic
RNA binding
endoribonuclease activity
protein binding
5'-3' exonuclease activity
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000558.hg.1 | ENST00000238112.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000558.hg.1 | ENST00000427001.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000558.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000558.hg.1 | ENST00000540142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000558.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02000558.hg.1 | TC02000047.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200145476.hg.1 | TC0200006647.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000238112.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000460593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000475482.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | ENST00000489629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200006647.hg.1 | NM_016207.3 | Refseq |

CPSF3
Genomic Map