Assay Details
| Gene Symbol: SEMA3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290061.1 | NP_001276990.1 | 8 - 9 | 1007 | 64 | |
| GenBank mRNA | AB083186.1 | - | 8 - 9 | 954 | 64 | |
More Information
Set Membership: |
|

SEMA3B
Genomic Map