| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014641.hg.1 |
ENST00000248244.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014641.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014641.hg.1 |
TC19001071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014642.hg.1 |
ENST00000248244.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014642.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014642.hg.1 |
TC19001071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014640.hg.1 |
ENST00000248244.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014640.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014640.hg.1 |
TC19001071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014643.hg.1 |
ENST00000248244.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014643.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19014643.hg.1 |
TC19001071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000621756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
ENST00000621756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
ENST00000621756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
TC1900009368.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
ENST00000621756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174895.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174900.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174900.hg.1 |
TC1900009368.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174900.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174896.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174896.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174896.hg.1 |
TC1900009368.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174896.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174896.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000621756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
ENST00000621756.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009368.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174898.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174898.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174898.hg.1 |
TC1900009368.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174898.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174898.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174899.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174899.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174899.hg.1 |
TC1900009368.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174899.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174899.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174897.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174897.hg.1 |
NM_182919.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900174897.hg.1 |
TC1900009368.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900174897.hg.1 |
ENST00000248244.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900174897.hg.1 |
NM_182919.3 |
Refseq |