Assay Details
| Gene Symbol: ODAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017855.3 | NP_060325.3 | 4 - 5 | 395 | 70 | |
| XM_005265698.3 | XP_005265755.1 | 5 - 6 | 459 | 70 | ||
| XM_006714245.3 | XP_006714308.1 | 5 - 6 | 436 | 70 | ||
| XM_017008331.1 | XP_016863820.1 | 3 - 4 | 351 | 70 | ||
| GenBank mRNA | AK000520.1 | - | 1 - 2 | 46 | 70 | |
| BG194365.1 | - | 4 - 5 | 398 | 70 | ||
| EF113908.1 | - | 4 - 5 | 397 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000380.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK000520.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
inflammatory response
positive regulation of gene expression
biomineral tissue development
regulation of actin cytoskeleton organization
odontogenesis of dentin-containing tooth
positive regulation of GTPase activity
cellular protein metabolic process
positive regulation of epithelial cell proliferation involved in wound healing
protein binding
positive regulation of protein phosphorylation
inflammatory response
positive regulation of gene expression
biomineral tissue development
regulation of actin cytoskeleton organization
odontogenesis of dentin-containing tooth
positive regulation of GTPase activity
cellular protein metabolic process
positive regulation of epithelial cell proliferation involved in wound healing
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005181.hg.1 | ENST00000396094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005181.hg.1 | ENST00000510709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005181.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005181.hg.1 | TC04000380.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005183.hg.1 | ENST00000396094.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005183.hg.1 | ENST00000510709.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005183.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005183.hg.1 | TC04000380.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400007775.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400153045.hg.1 | TC0400007775.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000396094.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000506248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000510709.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000510847.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | ENST00000514097.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | NM_017855.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400153047.hg.1 | TC0400007775.hg.1 | Clariom D TC |

ODAM
Genomic Map